Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Bright Classroom Activity Bowls
SDPSCI_ED

Catalog No. S13209
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13209 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13209 Supplier Roylco™ Supplier No. R55193
Add to Cart
Add to Cart

Colorful, bright classroom activity bowls that are handy for a variety of useful purposes.

  • Just the right size for little fingers to grab easily
  • Fun, bright colors are ideal for sorting craft supplies, or counting beads
  • Reusable bowls are great for keeping paints, glitter, glue or other messy materials neat and tidy
  • Top rack dishwasher safe and stackable, making clean-up a cinch
  • Assorted colors

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Age ≥3 Years
Dimensions (L x W) 5 x 15 cm (2 x 6 in.)
Includes 10 Colorful Bowls
Quantity 10 Bowls/Pk.
Weight (English) 1 lb.
Class Size 10 to 40 Students
Product Type Build Anything Bin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.