Learn More
Electron Microscopy Sciences Bright-Line Chamber Set
SDP
Catalog No. 5026894
Encompass_Preferred
Designed by Leica for accurate counting of blood cells platelets, dust/yeast cluster, and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm cubed/mm squared and 0.00025 mm squared per each square. Cover glass, pipettes.
Catalog No. 50-268-94 Supplier Electron Microscopy Sciences Supplier No. 6351410
Designed by Leica for accurate counting of blood cells platelets, dust/yeast cluster, and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm cubed/mm squared and 0.00025 mm squared per each square. Cover glass, pipettes.