Learn More
Hausser Scientific™ Bright-Line™ Counting Chamber PROMO
Catalog No. 0267151BS24
Change view
Click to view available options
Bright-Line Counting Chamber; Material: Milled Soda Lime Glass
Catalog No.
02-671-51BS24 Bright-Line Counting Chamber; Material: Milled Soda Lime Glass
1 options
This item is not returnable. View return policy
Catalog No. 02-671-51BS24 Supplier Hausser Scientific™ Supplier No. 0267151BS24
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Diamond-etched counting chamber with double improved Neubauer ruling.
  • One piece construction, ensuring long term durability and accuracy
  • 'V' slash at the loading side of each plateau facilitates charing and reduces the possibility of overflow into the moat
  • Each plateau features enhanced Neubauer rulings
  • Ruled surface is 0.1 mm below the cover glass (sold separately), limiting the volume of blood or fluid over a square mm at 0.1 mm3 and over each of 400 squares (within the central square mm) to 0.00025 mm3
  • Contact of the flat, polished cover glass surfaces with cover glass supports produces an exact volume of fluid over the counting area
  • Difference in surface tension characteristics between the metallic surface on the chamber and the polished cover glass assures smooth capillarity for precise loading and more even cell distribution
Specifications
Product Type Hemacytometer
Grid Type Bright Line
Capacity (Metric) 0.1 μL
Material Milled Soda Lime Glass
For Use With (Application) Double Improved Neubauer Ruling