Learn More
Sigma Aldrich Fine Chemicals Biosciences Bright-LineTM Hemacytomete1EA
Catalog No. 502904010
Encompass
The Bright-Line(TM) Hemacytometer is molded from a single piece of thermal and shock-resistant glass. An H-shaped moat forms two counting areas or plateaus. A V slash at the loading side of each plateau facilitates charing and reduces the possibility of overflow into the moat. Each plateau features enhanced Neubauer rulings.The ruled surface is 0.1mm below the cover glass limiting the volume of blood or fluid over a square mm at 0.1cu.mm and over each of 400 squares (within the central square mm) to 0.00025 cu. mm. Contact of the flat polished cover glass surfaces with cover glass supports produces an exact volume of fluid over the counting area. The difference in surface tension characteristics between the metallic surface on the chamber and the polished cover glass assures smooth capillarity for precise loading and more even cell distribution.Replacement cover slips sold separately Z375357
Catalog No. 50-290-4010 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3596291EA
The Bright-Line(TM) Hemacytometer is molded from a single piece of thermal and shock-resistant glass. An H-shaped moat forms two counting areas or plateaus. A V slash at the loading side of each plateau facilitates charing and reduces the possibility of overflow into the moat. Each plateau features enhanced Neubauer rulings.The ruled surface is 0.1mm below the cover glass limiting the volume of blood or fluid over a square mm at 0.1cu.mm and over each of 400 squares (within the central square mm) to 0.00025 cu. mm. Contact of the flat polished cover glass surfaces with cover glass supports produces an exact volume of fluid over the counting area. The difference in surface tension characteristics between the metallic surface on the chamber and the polished cover glass assures smooth capillarity for precise loading and more even cell distribution.Replacement cover slips sold separately Z375357