Learn More
Research Products International Corp BrightMax Film, Non-S, 50/pk
SDP
Catalog No. 502230154
Encompass
BrightMax sealing films are 203μm thick white vinyl films with 37μm acrylic adhesive for use with white microplates. The light-reflective properties of BrightMax plate-sealing films on clear-bottom plates maximize sensitivity of bioluminescence and chemiluminscence assays in bottom-reading luminometers. Sealing films may be applied to either the top or bottom of a plate. A split backing aids in applying the sealing film properly. White film can be used as backing matrix for microscopy of punchout samples from filter plates. BrightMax is a Trademark of Excel Scientific, Inc.
White light reflecting for white plates
Maximize intensity and signal-to-noise ratio
Withstand -40°C to +80°C
Catalog No. 50-223-0154 Supplier Research Products International Corp Supplier No. 202512
BrightMax sealing films are 203μm thick white vinyl films with 37μm acrylic adhesive for use with white microplates. The light-reflective properties of BrightMax plate-sealing films on clear-bottom plates maximize sensitivity of bioluminescence and chemiluminscence assays in bottom-reading luminometers. Sealing films may be applied to either the top or bottom of a plate. A split backing aids in applying the sealing film properly. White film can be used as backing matrix for microscopy of punchout samples from filter plates. BrightMax is a Trademark of Excel Scientific, Inc.
White light reflecting for white plates
Maximize intensity and signal-to-noise ratio
Withstand -40°C to +80°C