Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Roll

Catalog No. 19103107 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-103-107 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-107 Supplier Brady™ Supplier No. CH15P
  • $369.00
    $257.65 / Each of 1

    Save $111.35

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Rolls are surfactant treated used on wide range of chemicals.

  • Made of MAXX technology
  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Medium weight - optimal for most general maintenance needs

Specifications

Certifications/Compliance EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Color Black/Red on Yellow
Description Does Not Contain Recycled Content
Design Perforated
Capacity (English) 20 gal.
Capacity (Metric) 75.7 L
Material Polypropylene
Type Absorbent Roll
Packaging Quantity 30/Cs.
Thickness 1-ply
Weight Capacity Medium-weight
Dimensions (L x W) 150 ft. x 15 in.
Length (English) 150 ft.
Width (English) 15 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.