Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BrightSorb™ High Visibility Pillows

Catalog No. 19000677 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
18 x 18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-000-677 18 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-677 Supplier Brady™ Supplier No. CH1818

Ideal for extra protection during packaging and transport

  • Surfactant-treated polypropylene pillows can be used on a wide range of chemicals, acids and bases
  • They are chemically inert, so they will not react with aggressive fluids
  • Color and print help prevent risk of slips, trips and falls 

Specifications

Color Black/Red on Yellow
Capacity (English) 14 gal.
Material Surfactant-treated Polypropylene
Type SPC Absorbent
For Use With (Application) Absorbing chemicals, acids and bases
Dimensions (L x W) 18 x 18 in.
Length (English) 18 in.
Length (Metric) 20.32 cm
Width (Metric) 18 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.