Learn More
Alkali Scientific BrightStar AP, Alkaline Phosphatase Substrate, 1 x 100mL
SDP
Catalog No. 503028176
Encompass_Preferred
BrightStar AP is a sensitive, chemiluminescent substrate for alkaline phosphatase that allows for the rapid, reproducible detection of alkaline phosphatase-labeled molecules. BrightStar AP is supplied as a 0.25 mM ready-to-use aqueous solution, with no dilution necessary, for use in a variety of membrane-based applications, including Northern, Southern, and colony lift blots. BrightStar AP functions on both neutrally-charged and positively-charged membrane, giving the reagent added application versatility.
Catalog No. 50-302-8176 Supplier Alkali Scientific Supplier No. XR68
BrightStar AP is a sensitive, chemiluminescent substrate for alkaline phosphatase that allows for the rapid, reproducible detection of alkaline phosphatase-labeled molecules. BrightStar AP is supplied as a 0.25 mM ready-to-use aqueous solution, with no dilution necessary, for use in a variety of membrane-based applications, including Northern, Southern, and colony lift blots. BrightStar AP functions on both neutrally-charged and positively-charged membrane, giving the reagent added application versatility.