Learn More
Alkali Scientific BrightStar™ Femto HRP Chemiluminescent 2-Component Substrate ECL Kit, (7.5mL Luminol and 7.5mL Peroxide) 15 mL Total
SDP
Catalog No. 502388711
Encompass_Preferred
The BrightStar™ Femto HRP Chemiluminescent Substrate is a highly sensitive Luminol-based chemiluminescent substrate that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a two component kit system for extended shelf-life. Visualization may be carried out through x-ray film or a CCD camera. This is a 15mL kit of 7.5mL luminol reagent and 7.5mL peroxide solution.
Catalog No. 50-238-8711 Supplier Alkali Scientific Supplier No. XR93
The BrightStar™ Femto HRP Chemiluminescent Substrate is a highly sensitive Luminol-based chemiluminescent substrate that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a two component kit system for extended shelf-life. Visualization may be carried out through x-ray film or a CCD camera. This is a 15mL kit of 7.5mL luminol reagent and 7.5mL peroxide solution.