Learn More
Alkali Scientific BrightStar™ Nitrocellulose Membrane, Pore Size 0.45µm, 300 X 3000 mm, 1 Roll
SDP
Supplier:  Alkali Scientific XR641
Encompass_Preferred
BrightStar™ 0.45µm Pore Size Nitrocellulose Membrane is perfect for Western, Northern, and Southern blotting of low molecular weight proteins and nucleic acids. It wets readily with water-based solutions and is easy to handle. Compatible with standard immunoblotting and nucleic acid detection methods. BrightStar™ Nitrocellulose Membrane offers superior binding capacity with no background interference. One roll - 300 x 3000mm (30cm wide x 3.0m length).
Catalog No. 50-238-9548