Learn More
Alkali Scientific BrightStar™ Duration HRP Chemiluminescent 2-Component Substrate ECL Kit, (250mL Luminol and 250mL Peroxide) 500mL Total
SDP
Catalog No. 502335944
Encompass_Preferred
The BrightStar™ Duration HRP Chemiluminescent Substrate is a sensitive and long lasting luminol-based chemiluminescent two solution kit that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a two component kit system for extended shelf-life. Visualization may be carried out through x-ray film or digital imaging system utilizing a CCD camera. This 500mL kit of 250mL luminol reagent and 250mL peroxide solution.
Catalog No. 50-233-5944 Supplier Alkali Scientific Supplier No. XR92
The BrightStar™ Duration HRP Chemiluminescent Substrate is a sensitive and long lasting luminol-based chemiluminescent two solution kit that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a two component kit system for extended shelf-life. Visualization may be carried out through x-ray film or digital imaging system utilizing a CCD camera. This 500mL kit of 250mL luminol reagent and 250mL peroxide solution.