Learn More
Alkali Scientific BrightStar™ Femto HRP Chemiluminescent Substrate, Pre-Mixed Single Component Luminol and Peroxide, ECL Mix, 100mL
SDP
Catalog No. 502336219
Encompass_Preferred
The BrightStar™ Femto HRP Chemiluminescent Substrate is a highly sensitive Luminol-based chemiluminescent substrate that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a single component mix system for added convenience. Visualization may be carried out through x-ray film or a CCD camera. This is a 100mL kit luminol reagent and peroxide solution.
Catalog No. 50-233-6219 Supplier Alkali Scientific Supplier No. XR96
The BrightStar™ Femto HRP Chemiluminescent Substrate is a highly sensitive Luminol-based chemiluminescent substrate that may be used for any blotting applications utilizing horseradish peroxidase (HRP)-conjugated antibodies. The HRP substrates are compatible with various blocking buffers and can be applied on PVDF or nitrocellulose with low backgrounds. The HRP Western substrates come as a single component mix system for added convenience. Visualization may be carried out through x-ray film or a CCD camera. This is a 100mL kit luminol reagent and peroxide solution.