Learn More
Medchemexpress LLC Brigimadlin (Synonyms: BI 907828) 10mg
SDP
Catalog No. NC3351711
Encompass_Preferred
Brigimadlin (BI 907828) is an orally active E3 ubiquitin-protein ligase MDM-2 inhibitor, preventing MDM-2 from negatively regulating the tumor suppressor p53. Brigimadlin can be used for antineoplastic research.
Catalog No. NC3351711 Supplier Medchemexpress LLC Supplier No. HY15285910MG
Brigimadlin (BI 907828) is an orally active E3 ubiquitin-protein ligase MDM-2 inhibitor, preventing MDM-2 from negatively regulating the tumor suppressor p53. Brigimadlin can be used for antineoplastic research.