Learn More
Bioworld Brij-35, 500 g
Catalog No. 50255644
Encompass_Preferred
Also known as Polyoxyethylene Lauryl EtherA non-ionic detergent suitable for Stein-Moore chromatographyHigh Purity GradeWater: <3.0%
Catalog No. 50-255-644 Supplier Bioworld Supplier No. 402300081
Also known as Polyoxyethylene Lauryl EtherA non-ionic detergent suitable for Stein-Moore chromatographyHigh Purity GradeWater: <3.0%