Learn More
Bioworld Brij-35, 1 kg
Catalog No. 50255645
Encompass_Preferred
Also known as Polyoxyethylene Lauryl EtherA non-ionic detergent suitable for Stein-Moore chromatographyHigh Purity GradeWater: <3.0%
Catalog No. 50-255-645 Supplier Bioworld Supplier No. 402300082
Also known as Polyoxyethylene Lauryl EtherA non-ionic detergent suitable for Stein-Moore chromatographyHigh Purity GradeWater: <3.0%