Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brij™-35 Surfact-Amps™ Detergent Solution

Catalog No. PI85117 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
6 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85117 50 mL
PI28316 6 x 10 mL
PI85118 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI85117 Supplier Thermo Scientific™ Supplier No. 85117
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.

Thermo Scientific™ Brij-35 Detergent Solutions are stabilized 10% and 30% solutions of Brij-35 detergent for use in various protein methods.

Additional Information and Online Ordering

Brij-35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or as a surfactant in various HPLC applications. The detergent is offered in two forms: a Thermo Scientific Surfact-Amps™ Detergent Solution (high-purity, 10% solution) and a standard grade 30% solution.

Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neet detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

  • Brij-35 – a nonionic detergent for use in various protein methods

  • Accurate – 10% and 30% detergent solutions in ultrapure water

  • Easy-to-use – solutions are simple to dispense and dilute for use

Detergent Properties:

  • Molecular Weight: 1225g
  • Detergent Class: Nonionic
  • Aggregation Number: 40
  • Micelle Molecular Weight: 49,000g
  • Critical Micelle Concentration (CMC): 0.09mM (0.011%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Surfact-Amps Detergent Solution (Part No. 28316):

  • Accurate – precise 10% detergent solution in ultrapure water

  • Easy-to-use – solution is simple to dispense and dilute for use

  • Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

  • Stable – packed in glass ampules under inert nitrogen gas

TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store at Room Temperature
Form Liquid
Product Line Brij, Surfact-Amps
Reagent Type Detergent Solution
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.