Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij™ 35, Fisher BioReagents

Catalog No. BP345500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP345-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP345-500

missing translation for 'validMainframeMsg'

This nonionic detergent is used for solubilizing proteins and isolating membrane complexes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene (23) Lauryl Ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color White
Physical Form Solid
Melting Point 40°C
pH 5.5 to 7.0
Quantity 500 g
Hydroxyl Value 40 to 60
Identification Pass Test
Packaging Glass Bottle
Water 3% max.
Linear Formula C12H25 (OCH2CH2)n
Merck Index 15, 7675
Formula Weight 1199.56
Chemical Name or Material Brij™ 35
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.