Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij™ 35, For Synthesis, MilliporeSigma™

Catalog No. M8019620250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8019620250 250 g Plastic bottle
M8019621000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M8019620250 Supplier MilliporeSigma Supplier No. 8.01962.0250
Add to Cart
Add to Cart
  • Used for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyethylene glycol lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color White
Physical Form Liquid
Boiling Point 100°C
Density 1.05g/cm3 (20°C)
pH 5.5 to 7.0 (at 100 g/L, H2O, 20°C)
Quantity 250 g
Flash Point >149°C
Hydroxyl Value 45 to 50
Packaging Plastic bottle
Pour Point 33°C
Water <3.0% (Karl Fisher)
Solubility Information Soluble in water at 25°C
Chemical Name or Material Polyoxyethylene lauryl ether
Show More Show Less
EINECSNumber 500-002-6
Recommended Storage Store below 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.