Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij™ C20, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
9004-95-9
C56H114O21
1123.507
MFCD00080892
NLMKTBGFQGKQEV-UHFFFAOYSA-N
Polyoxyethylene(20) cetyl ether
2724259
2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hexadecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
9004-95-9
C56H114O21
1123.507
MFCD00080892
NLMKTBGFQGKQEV-UHFFFAOYSA-N
Polyoxyethylene(20) cetyl ether
2724259
2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hexadecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
Acid Value 1mg KOH/g max.
Additional Information Without antioxidant
Dioxane free dioxane: < 10ppm
Ethylene Oxide free ethyleneoxide: < 1ppm
Hydroxyl Value 45 to 60mg KOH/g
Linear Formula C16H33(OCH2CH2)20OH
Infrared Spectrum Authentic
Merck Index 10, 7449
Packaging Glass bottle
Formula Weight 1123.5
Physical Form Pellets
Solubility Solubility in water: soluble. Other solubilities: soluble in lower alcohols, insoluble in propylene glycol, coal-tar hydrocarbo, n, cottonseed and mineral oil
Water 1% max.
Color White
Melting Point 35.4°C to 39.6°C
Flash Point >149°C
Quantity 5g
Chemical Name or Material Brij™ C20
Show More Show Less

RTECSNumber : TR1581470

TSCA : TSCA