Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Brij™ 93

Catalog No. 388866100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-886-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-886-6100ML Supplier Sigma Aldrich Supplier No. 388866100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Brij(R) 93 is a non-ionic surfactant which has a polar group in the form of polyoxyethylene that is separated by the polyethylene chains. Its molecular structure includes polyethylene glycol (PEG) single chain. It is slightly soluble in water and has a hydrophobic nature.Brij is a registered trademark of Croda International PLC

Specifications

CAS 9004-98-2
Density 0.912 g/mL (at 25°C)
Molecular Formula C22H44O3
Refractive Index n20/D 1.462 (literature)
Linear Formula C18H35(OCH2CH2)nOH, n ~2
Quantity 100 mL
MDL Number MFCD00074369
Synonym Polyethylene glycol oleyl ether; Polyoxyethylene (2) oleyl ether
Molecular Weight (g/mol) 356.58
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.