Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij(R) L23, Spectrum™ Chemical
SDP

Catalog No. 18620355 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-355 12 kg
18-620-353 500 g
18-620-354 2.5 kg
18-620-352 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-355 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BR10512KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9002-92-0

Specifications

pH Slightly Acidic
Quantity 12 kg
Packaging Poly Bottle
Chemical Name or Material Brij L23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.