Learn More
Medchemexpress LLC Brilacidin (tetrahydrochloride) | 1224095-99-1 | 1082.75 | 5 MG
SDP
Catalog No. 5000401008
Encompass_Preferred
Brilacidin tetrahydrochloride is an anti-infective antimicrobial. It demonstrates MIC90s of 1 and 8 μg/mL for Gram-positive bacteria such as *Streptococcus pneumonia* and *Streptococcus viridans*, and MIC90s of 8 and 4 μg/mL for Gram-negative bacteria including *Haemophilus influenza* and *Pseudomonas aeruginosa*. This compound acts as a defensin mimetic antibiotic.
  • Functions as an anti-infective antimicrobial
  • Acts as a defensin mimetic antibiotic compound
Catalog No. 50-004-01008 Supplier Medchemexpress LLC Supplier No. HY19892A5MG
Brilacidin tetrahydrochloride is an anti-infective antimicrobial. It demonstrates MIC90s of 1 and 8 μg/mL for Gram-positive bacteria such as *Streptococcus pneumonia* and *Streptococcus viridans*, and MIC90s of 8 and 4 μg/mL for Gram-negative bacteria including *Haemophilus influenza* and *Pseudomonas aeruginosa*. This compound acts as a defensin mimetic antibiotic.
  • Functions as an anti-infective antimicrobial
  • Acts as a defensin mimetic antibiotic compound