Learn More
Medchemexpress LLC Brilacidin tetrahydrochloride | 1224095-99-1 | 99.10% | 1082.75 | 100 MG
SDP
Catalog No. 5000401630
Encompass_Preferred
Brilacidin tetrahydrochloride is an anti-infective antimicrobial compound with demonstrated activity against both Gram-positive and Gram-negative bacteria. It is a defensin mimetic antibiotic that primarily targets the bacterial cell membrane through depolarization. It shows lower minimum inhibitory concentrations for Staphylococcus aureus and Staphylococcus epidermidis compared to other bacterial groups. For research use only.
  • Anti-infective antimicrobial with broad-spectrum activity.
  • Effective against Gram-positive bacteria including Streptococcus pneumonia and Streptococcus viridans.
  • Also active against Gram-negative bacteria such as Haemophilus influenza and Pseudomonas aeruginosa.
  • Functions as a defensin mimetic antibiotic.
  • Primary mechanism involves bacterial cell membrane depolarization.
Catalog No. 50-004-01630 Supplier Medchemexpress LLC Supplier No. HY19892A100MG
Brilacidin tetrahydrochloride is an anti-infective antimicrobial compound with demonstrated activity against both Gram-positive and Gram-negative bacteria. It is a defensin mimetic antibiotic that primarily targets the bacterial cell membrane through depolarization. It shows lower minimum inhibitory concentrations for Staphylococcus aureus and Staphylococcus epidermidis compared to other bacterial groups. For research use only.
  • Anti-infective antimicrobial with broad-spectrum activity.
  • Effective against Gram-positive bacteria including Streptococcus pneumonia and Streptococcus viridans.
  • Also active against Gram-negative bacteria such as Haemophilus influenza and Pseudomonas aeruginosa.
  • Functions as a defensin mimetic antibiotic.
  • Primary mechanism involves bacterial cell membrane depolarization.