Learn More
Medchemexpress LLC Brilaroxazine | 1239729-06-6 | 98.8% | 450.36 g/mol | C22H25Cl2N3O3 | 5 MG
SDP
Catalog No. 5000264438
Encompass_Preferred
Brilaroxazine (RP5063) is an investigational atypical antipsychotic and serotonin-dopamine system modulator supplied for research use. It is offered as a purified solid and as a DMSO solution, intended for preclinical pharmacology, receptor profiling, and in vitro studies.
  • Investigational atypical antipsychotic and dopamine/5-HT modulator.
  • High purity (98.8%).
  • Chemical formula C22H25Cl2N3O3; molecular weight 450.36 g/mol.
  • Available as solid and as a 10 mM solution in DMSO.
  • Suitable for receptor binding, pharmacology, and preclinical studies.
  • Offered in small research pack sizes, including 5 mg.
Catalog No. 50-002-64438 Supplier Medchemexpress LLC Supplier No. HY1091125MG
Brilaroxazine (RP5063) is an investigational atypical antipsychotic and serotonin-dopamine system modulator supplied for research use. It is offered as a purified solid and as a DMSO solution, intended for preclinical pharmacology, receptor profiling, and in vitro studies.
  • Investigational atypical antipsychotic and dopamine/5-HT modulator.
  • High purity (98.8%).
  • Chemical formula C22H25Cl2N3O3; molecular weight 450.36 g/mol.
  • Available as solid and as a 10 mM solution in DMSO.
  • Suitable for receptor binding, pharmacology, and preclinical studies.
  • Offered in small research pack sizes, including 5 mg.