Learn More
Cayman Chemical BrIlInt Black BN 10g
SDP
Catalog No. 502950276
Encompass_Preferred
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)
Catalog No. 50-295-0276 Supplier Cayman Chemical Supplier No. 3446210 G
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)