Learn More
Cayman Chemical Brilliant Black BN, Purity: ≥60%, CAS: 2519-30-4, 25 g
SDP
Catalog No. 502950285
Encompass_Preferred
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)
Catalog No. 50-295-0285 Supplier Cayman Chemical Supplier No. 3446225 G
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)