Learn More
Cayman Chemical BrIlInt Black BN 50g
SDP
Catalog No. 502949791
Encompass_Preferred
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)
Catalog No. 50-294-9791 Supplier Cayman Chemical Supplier No. 3446250 G
A sulfonated azo dye with antiviral activity; reduces the infectivity of EV71 in rhabdomyosarcoma cells (IC50 = 10.1 µM)