Learn More
Cayman Chemical Brilliant Blue G, CAS: 6104-58-1, 25 g
SDP
Catalog No. 502954021
Encompass_Preferred
A dye which stains proteins in gels with ease and sensitivity; used in the Bradford to quantify the amount of protein in a solution; selectively inhibits P2X7 (IC50s = 10.1 nM and 265 nM for rat and human P2X7, respectively) and reduces neurological injury secondary to traumatic brain injury in mice
Catalog No. 50-295-4021 Supplier Cayman Chemical Supplier No. 1432025 G
A dye which stains proteins in gels with ease and sensitivity; used in the Bradford to quantify the amount of protein in a solution; selectively inhibits P2X7 (IC50s = 10.1 nM and 265 nM for rat and human P2X7, respectively) and reduces neurological injury secondary to traumatic brain injury in mice