Learn More
Biolegend Brilliant Violet 605™ anti-human CD8
Supplier:  Biolegend 344741
Encompass_Preferred
Brilliant Violet 605™ anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 25 tests
Catalog No. NC1024775