Learn More
Medchemexpress LLC Brilliant black BN | 2519-30-4 | 50 MG
SDP
Catalog No. 5000341199
Encompass_Preferred
Brilliant black BN (E151) is an azo dye and a food colorant. It is a promising antiviral agent against EV71 infection, inhibiting the interaction between EV71 and its cellular uncoating factor cyclophilin A. This substance has potential for the investigation of contagious diseases and is intended for research use only.
  • Inhibits EV71 infection in a dose-dependent manner
  • Effective against 28 strains of EV71, CVA16, and CVA6
  • Provides protective effects in EV71 challenged AG129 mice
  • Soluble in DMSO at 125 mg/mL
Catalog No. 50-003-41199 Supplier Medchemexpress LLC Supplier No. HY12838250MG
Brilliant black BN (E151) is an azo dye and a food colorant. It is a promising antiviral agent against EV71 infection, inhibiting the interaction between EV71 and its cellular uncoating factor cyclophilin A. This substance has potential for the investigation of contagious diseases and is intended for research use only.
  • Inhibits EV71 infection in a dose-dependent manner
  • Effective against 28 strains of EV71, CVA16, and CVA6
  • Provides protective effects in EV71 challenged AG129 mice
  • Soluble in DMSO at 125 mg/mL