Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Black, MP Biomedicals

Catalog No. icn202327 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202327 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202327 Supplier MP Biomedicals, Inc Supplier No. 0520232710
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2519-30-4
Molecular Formula C28H17N5Na4O14S4
Molecular Weight (g/mol) 867.664
InChI Key AGKKBBSOKGLVTM-YENKNCRGSA-J
PubChem CID 6321379
IUPAC Name tetrasodium;(6E)-4-acetamido-5-oxo-6-[[7-sulfonato-4-[(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]hydrazinylidene]naphthalene-1,7-disulfonate
SMILES CC(=O)NC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=C(C(=NNC3=C4C=C(C=CC4=C(C=C3)N=NC5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-])C2=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Formula Weight 871.74
Quantity 10 g
Chemical Name or Material Brilliant Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.