Learn More
Medchemexpress LLC Brilliant blue G-250 | 6104-58-1 | 78482 | >=95.0% | 250 MG
SDP
Catalog No. 5000413959
Encompass_Preferred
Brilliant Blue G-250 is a dye primarily used for visualizing proteins separated by SDS-PAGE, offering a straightforward staining procedure and high quantitation. It also functions as a safe and highly selective P2x7R antagonist, inactivating the NLRP3 inflammasome.
  • Determines protein concentrations by measuring absorbance at 595 nm in the Bradford protein assay.
  • Antagonist activity at human P2X7R with an IC50 of 1.37 μM in HEK293 cells.
  • Anti-inflammatory activity in LPS/IFNγ-differentiated peritoneal macrophages with an IC50 of 19.77 μM.
  • Anti-inflammatory activity in LPS/IFNγ-differentiated THP-1 cells with an IC50 of 19.27 μM.
Catalog No. 50-004-13959 Supplier Medchemexpress LLC Supplier No. 250MGHYD0014
Brilliant Blue G-250 is a dye primarily used for visualizing proteins separated by SDS-PAGE, offering a straightforward staining procedure and high quantitation. It also functions as a safe and highly selective P2x7R antagonist, inactivating the NLRP3 inflammasome.
  • Determines protein concentrations by measuring absorbance at 595 nm in the Bradford protein assay.
  • Antagonist activity at human P2X7R with an IC50 of 1.37 μM in HEK293 cells.
  • Anti-inflammatory activity in LPS/IFNγ-differentiated peritoneal macrophages with an IC50 of 19.77 μM.
  • Anti-inflammatory activity in LPS/IFNγ-differentiated THP-1 cells with an IC50 of 19.27 μM.