Learn More
Research Products International Corp Brilliant Blue R-250 [Coomassie Blue], 50 Grams
SDP
Catalog No. 501368121
Encompass_Preferred
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.
Catalog No. 50-136-8121 Supplier Research Products International Corp Supplier No. B4300050.0
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.