Learn More
Alkali Scientific Brilliant Blue R 250 [Coomassie Blue], 25 G
SDP
Catalog No. 5000660237
Encompass_Preferred
Brilliant Blue R 250, also known as Coomassie Blue, is a commonly used protein-staining dye. It is widely used in laboratory procedures like electrophoresis and western blotting to stain proteins in gels. This 25g container provides an ample quantity for routine laboratory use, allowing consistent staining for protein detection in SDS-PAGE experiments. The dye binds to proteins, providing clear visualization of protein bands, which is essential for molecular biology applications. Coomassie Blue R 250 is known for its high sensitivity and is often used in conjunction with protein assays to quantify protein concentration in biological samples. It can be used with various detection methods, including imaging and spectrophotometry.
Catalog No. 50-006-60237 Supplier Alkali Scientific Supplier No. BRI001CP
Brilliant Blue R 250, also known as Coomassie Blue, is a commonly used protein-staining dye. It is widely used in laboratory procedures like electrophoresis and western blotting to stain proteins in gels. This 25g container provides an ample quantity for routine laboratory use, allowing consistent staining for protein detection in SDS-PAGE experiments. The dye binds to proteins, providing clear visualization of protein bands, which is essential for molecular biology applications. Coomassie Blue R 250 is known for its high sensitivity and is often used in conjunction with protein assays to quantify protein concentration in biological samples. It can be used with various detection methods, including imaging and spectrophotometry.