Learn More
Alkali Scientific Brilliant Blue R 250 [Coomassie Blue], 250 Grams
SDP
Catalog No. 5000660238
Encompass_Preferred
Brilliant Blue R 250 (Coomassie Blue) is a protein-staining dye available in a 250g format for large-scale applications. It is used extensively in protein electrophoresis, particularly in SDS-PAGE, where it stains proteins in the gel matrix for easy visualization and analysis. The dye binds to proteins, allowing researchers to detect and quantify protein bands with high sensitivity. This 250g container is ideal for high-throughput laboratories or for researchers conducting large-scale protein analysis. Coomassie Blue is widely used in biochemistry and molecular biology labs for protein quantification, protein gel analysis, and other molecular techniques.
Catalog No. 50-006-60238 Supplier Alkali Scientific Supplier No. BRI003CP
Brilliant Blue R 250 (Coomassie Blue) is a protein-staining dye available in a 250g format for large-scale applications. It is used extensively in protein electrophoresis, particularly in SDS-PAGE, where it stains proteins in the gel matrix for easy visualization and analysis. The dye binds to proteins, allowing researchers to detect and quantify protein bands with high sensitivity. This 250g container is ideal for high-throughput laboratories or for researchers conducting large-scale protein analysis. Coomassie Blue is widely used in biochemistry and molecular biology labs for protein quantification, protein gel analysis, and other molecular techniques.