Learn More
Alkali Scientific Brilliant Blue R 250 [Coomassie Blue], 50 G
Supplier: Alkali Scientific BRI002CP
Brilliant Blue R 250 (Coomassie Blue) is a popular protein-staining dye used in laboratory applications such as SDS-PAGE and western blotting. This 50g container offers a larger quantity, making it ideal for high-throughput labs or extensive protein analysis. Coomassie Blue binds to proteins in a gel matrix, allowing researchers to visualize and quantify protein bands with high precision. The dye is widely used for protein separation and analysis, particularly in molecular biology, biochemistry, and clinical laboratories. Its strong staining properties and ease of use make it a go-to choice for protein gel electrophoresis, enabling effective protein detection and quantification for a variety of applications.