Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Blue R Staining Solution, Sigma-Aldrich

Catalog No. 01258994 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-994 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-994 Supplier MilliporeSigma Supelco Supplier No. B65291L
Add to Cart
Add to Cart

Brilliant Blue R staining solution is especially designed for use in staining protein in agarose gels following immunoelectrophoresis and Ouchterlony gels. The stain contains ethanol and acetic acid so gels do not require fixing prior to staining. The immunoelectrophoresis or Ouchterlony gel is first washed with water and saline to remove non-precipitated proteins and then dried. The gel is then immersed in staining solution for 30 min and destained with 10% acetic acid.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brilliant Blue R Staining Solution
CAS 6104-59-2
Molecular Weight (g/mol) 825.97 g/mol
Color Dark Blue
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.