Learn More
ION BIOSCIENCES Brilliant Calcium Assay Flex kit - 10 plates
Catalog No. NC4043362
Encompass_Preferred
ION Biosciences’ Brilliant Calcium (Ca²⁺) Assay Flex kit is a total assay solution for multi-well plate-based, high-throughput measurements of dynamic changes in intracellular Ca²⁺. The ION Biosciences’ Brilliant Calcium Assay is useful for a wide range of effectors of intracellular Ca²⁺ including many G protein-coupled receptors (GPCRs), divalent cation channels, and other proteins capable of transporting Ca²⁺ or affecting intracellular Ca²⁺ dynamics. In multi-well, plate-based formats, the Brilliant Calcium Assay can be used to discover and characterize the effects of tens to many tens-of-thousands of compounds and environmental factors which can act to affect intracellular Ca²⁺ levels. 10-plate kit.
Catalog No. NC4043362 Supplier ION BIOSCIENCES Supplier No. 1000010
ION Biosciences’ Brilliant Calcium (Ca²⁺) Assay Flex kit is a total assay solution for multi-well plate-based, high-throughput measurements of dynamic changes in intracellular Ca²⁺. The ION Biosciences’ Brilliant Calcium Assay is useful for a wide range of effectors of intracellular Ca²⁺ including many G protein-coupled receptors (GPCRs), divalent cation channels, and other proteins capable of transporting Ca²⁺ or affecting intracellular Ca²⁺ dynamics. In multi-well, plate-based formats, the Brilliant Calcium Assay can be used to discover and characterize the effects of tens to many tens-of-thousands of compounds and environmental factors which can act to affect intracellular Ca²⁺ levels. 10-plate kit.