Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Brilliant Cresyl Blue, Certified, C.I. DcV-5 10 GR
SDP

Catalog No. 50980212
Encompass_Preferred

(Brilliant Blue C; Cresyl Blue 2RN)
CAS #10127-36-3 C₁₇H₂₁N₄OCl F.W. 332.84
Solubility: 3.0% Water; 2.6% Alcohol; 2.25% Cellosolve; 8.0% Glycol; 9.5% Xylene

Platelets & reticulum of immature red cells. Counterstained with Wright's stain.

Catalog No. 50-980-212 Supplier Electron Microscopy Sciences Supplier No. 11465
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Brilliant Blue C; Cresyl Blue 2RN)
CAS #10127-36-3 C₁₇H₂₁N₄OCl F.W. 332.84
Solubility: 3.0% Water; 2.6% Alcohol; 2.25% Cellosolve; 8.0% Glycol; 9.5% Xylene

Platelets & reticulum of immature red cells. Counterstained with Wright's stain.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.