Learn More
Brilliant Cresyl Blue Solution, 100 mL, G-Biosciences

Description
- The dye stains the nucleic acids in the reticulocytes, thus morphologically distinguishes reticulocytes from erythrocytes.
Specifications
Specifications
| Chemical Name or Material | Brilliant Blue C |
| CAS | 81029-05-2 |
| Color | Blue to Dark Blue |
| pH | 5.0 to 7.0 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 385.96 |
| Percent Purity | 98% |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.