Learn More
Brilliant Cresyl Blue Stain, 1% (w/v) in 0.85% (w/v) Sodium Chloride, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Brilliant Cresyl Blue Stain |
| CAS | 7732-18-5, 81029-05-2, 7647-14-5 |
| CAS Min % | 96.0, 0.96, 0.82 |
| CAS Max % | 100.0, 1.0, 0.85 |
| Concentration | 1% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | 0.85% (w/v) Sodium Chloride |
| Color | Blue/Purple |
| pH | 4.4 |
| Quantity | 120 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.