Learn More
Description
Brilliant Green is a dye and an antiseptic agent.
Specifications
Specifications
| Chemical Name or Material | Brilliant Green |
| CAS | 633-03-4 |
| Color | Blue/Green |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 482.63 g/mol |
| Percent Purity | ≥85% |
| Grade | Certified Biological Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.