Learn More
MOLTOX Molecular Toxicology Inc Brilliant Green Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948815
Encompass_Preferred
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S. typhi from materials such as feces. It is used in procedures for the examination of water and wastewater as well as for use in the performance of Microbial Limit Tests. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-815 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S015
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S. typhi from materials such as feces. It is used in procedures for the examination of water and wastewater as well as for use in the performance of Microbial Limit Tests. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.