Learn More
NEOGEN Brilliant Green Agar, 500 g
SDP
Catalog No. 502015939
Encompass_Preferred
Brilliant Green Agar is used for the selective isolation of Salmonella spp. in a laboratory setting. Brilliant Green Agar is not intended for use in the diagnosis of disease or other conditions in humans.Kristensen, Lester, and Jurgens first described the use of Brilliant Green Agar as a primary plating medium for the isolation of Salmonella. The report described the medium as useful for the differentiation of “paratyphoid B” from other intestinal gram-negative bacilli. Kaufmann modified their formula and used Brilliant Green Agar in addition to Tetrathionate Broth for the isolation of Salmonella from stool specimens. The outstanding selectivity of this medium permits use of moderately heavy inocula, which should be evenly distributed over the surface. Brilliant Green Agar is used in the microbial limits test. Brilliant Green Agar supplemented with novobiocin is used in food testing.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-agar-ncm0283
Catalog No. 50-201-5939 Supplier NEOGEN Supplier No. 700003660
Brilliant Green Agar is used for the selective isolation of Salmonella spp. in a laboratory setting. Brilliant Green Agar is not intended for use in the diagnosis of disease or other conditions in humans.Kristensen, Lester, and Jurgens first described the use of Brilliant Green Agar as a primary plating medium for the isolation of Salmonella. The report described the medium as useful for the differentiation of “paratyphoid B” from other intestinal gram-negative bacilli. Kaufmann modified their formula and used Brilliant Green Agar in addition to Tetrathionate Broth for the isolation of Salmonella from stool specimens. The outstanding selectivity of this medium permits use of moderately heavy inocula, which should be evenly distributed over the surface. Brilliant Green Agar is used in the microbial limits test. Brilliant Green Agar supplemented with novobiocin is used in food testing.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-agar-ncm0283