Learn More
Bioworld Brilliant Green Agar, 500 g
Catalog No. 50254455
Encompass_Preferred
Brilliant Green AgarUsed in the isolation of Salmonella species from foods, especially eggspH: 7.0Dehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange-brown agarSoluble in boiling waterComposition:Yeast Extract Pancreatic Digest of Casein Peptone Peptic Digest of Animal Tissue Sodium Chloride Lactose Sucrose Brillant Green Phenol Red Agar
Catalog No. 50-254-455 Supplier Bioworld Supplier No. 306230001
Brilliant Green AgarUsed in the isolation of Salmonella species from foods, especially eggspH: 7.0Dehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange-brown agarSoluble in boiling waterComposition:Yeast Extract Pancreatic Digest of Casein Peptone Peptic Digest of Animal Tissue Sodium Chloride Lactose Sucrose Brillant Green Phenol Red Agar