Learn More
Bioworld Brilliant Green Agar, 1 kg
Catalog No. 50254456
Encompass_Preferred
Brilliant Green AgarUsed in the isolation of Salmonella species from foods, especially eggspH: 7.0Dehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange-brown agarSoluble in boiling waterComposition:Yeast Extract Pancreatic Digest of Casein Peptone Peptic Digest of Animal Tissue Sodium Chloride Lactose Sucrose Brillant Green Phenol Red Agar
Catalog No. 50-254-456 Supplier Bioworld Supplier No. 306230002
Brilliant Green AgarUsed in the isolation of Salmonella species from foods, especially eggspH: 7.0Dehydrated Appearance: pink, homogeneous powderPrepared Appearance: orange-brown agarSoluble in boiling waterComposition:Yeast Extract Pancreatic Digest of Casein Peptone Peptic Digest of Animal Tissue Sodium Chloride Lactose Sucrose Brillant Green Phenol Red Agar