Learn More
NEOGEN Brilliant Green Agar (Modified), 500 g
SDP
Catalog No. 502015321
Encompass_Preferred
Brilliant Green Agar is used for the selection and differentiation of Salmonella (other than S. typhi) from foods or feedstuffs. Brilliant Green Agar is not intended for use in the diagnosis of disease or other conditions in humans. First introduced by Kristensen et al in 1925 as a selective medium for the isolation of Salmonella (except S. typhi). The medium was modified by the Netherlands Institute for Public Health, Utrecht. The modification was to increase the selectivity of the medium by increasing the dye concentration. This formulation is quoted by the International Standards Organization, standard European Community Methods, the American Public Health Association and the Association of Official Analytical Chemists. The medium is suitable for subcultures from selective enrichment media. However, because this medium is highly selective, small numbers of salmonellae may be missed.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-agar
Catalog No. 50-201-5321 Supplier NEOGEN Supplier No. 700004439
Brilliant Green Agar is used for the selection and differentiation of Salmonella (other than S. typhi) from foods or feedstuffs. Brilliant Green Agar is not intended for use in the diagnosis of disease or other conditions in humans. First introduced by Kristensen et al in 1925 as a selective medium for the isolation of Salmonella (except S. typhi). The medium was modified by the Netherlands Institute for Public Health, Utrecht. The modification was to increase the selectivity of the medium by increasing the dye concentration. This formulation is quoted by the International Standards Organization, standard European Community Methods, the American Public Health Association and the Association of Official Analytical Chemists. The medium is suitable for subcultures from selective enrichment media. However, because this medium is highly selective, small numbers of salmonellae may be missed.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-agar