Learn More
NEOGEN Brilliant Green Bile 2% Broth, 5 kg
SDP
Catalog No. 502015292
Encompass_Preferred
Brilliant Green Bile Broth 2% is used for the detection of coliform bacteria in water, food, and dairy products in a laboratory setting. Brilliant Green Bile Broth 2% is not intended for use in the diagnosis of disease or other conditions in humans.The coliform group of bacteria includes aerobic and facultative anaerobic, Gram-negative, non-sporeforming bacilli that ferment lactose and form acid and gas at 35°C within 48 hours. Members of the Enterobacteriacae comprise the majority of this group, but organisms such as Aeromonas spp. may also be included. Procedures to detect and confirm coliforms are used in testing water, foods, dairy products and other materials. Brilliant Green Bile Broth 2% is used to confirm a positive presumptive test result.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-bile-2-percent-broth
Catalog No. 50-201-5292 Supplier NEOGEN Supplier No. 700003090
Brilliant Green Bile Broth 2% is used for the detection of coliform bacteria in water, food, and dairy products in a laboratory setting. Brilliant Green Bile Broth 2% is not intended for use in the diagnosis of disease or other conditions in humans.The coliform group of bacteria includes aerobic and facultative anaerobic, Gram-negative, non-sporeforming bacilli that ferment lactose and form acid and gas at 35°C within 48 hours. Members of the Enterobacteriacae comprise the majority of this group, but organisms such as Aeromonas spp. may also be included. Procedures to detect and confirm coliforms are used in testing water, foods, dairy products and other materials. Brilliant Green Bile Broth 2% is used to confirm a positive presumptive test result.For more information visit https://www.neogen.com/solutions/Microbiology/brilliant-green-bile-2-percent-broth