Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Brilliant Green Bile Bottled Broth with Durham tubes 9mL vials

Catalog No. 10486163 Shop All GVS Products
Change view
Click to view available options
20 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
10-486-163 20 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-486-163 Supplier GVS™ Supplier No. 10496710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brilliant Green Bile Broth is used to detect coliforms in water, milk and other samples. Ready-to-use 9mL ampoules save time and money by shortening prep time and minimizing possible contamination. The 9mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

Description

  • BGBB contains two inhibitors of both gram-positive and selected gram-negative organisms, namely, oxgall and brilliant green dye. Fermentation is detected by gas production.

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 9 mL Portions, Reduce Risk of Cross-Contamination.
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Use with membrane filters to culture bacteria, yeasts, and molds.
  • For the detection of coliforms in water and other samples.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits.
This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Appearance Green
ATCC® 25922, 13048
Broth Type Brilliant Green Bile Bottled Broth
Certifications/Compliance Batch-specific quality certificate in each pack
Color Green
Country of Origin United States
Additive Bxgall, Brilliant Green Dye
Detects coliforms in water, milk and other samples.
Form Liquid
Format Vial
Manufacturing Origin United States
Media Type Brilliant Green Bile Bottled Broth
Preparation Method Ready-to-Use
Product Type Bacterial Growth Medium
Sterility Sterile
Target Organism Class Coliforms
Test Type Microbial Detection
Fill Volume 9 mL
Formulation includes oxgall and brilliant green dye.
For Use With (Application) Detection of Coliforms, Research of Food Products, Water, Clinical Samples
Includes Durham Tubes
No. per Pack 20
Packaging Type Vial
Quantity 9 mL
Specimen Type Water, Milk, Food
Protein Source Bovine Bile
Manufacturing Quality ISO 9001, ISO 13485, GMP, TR CU 004/2011
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.