Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant green, MP Biomedicals™

Catalog No. ICN15051680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15051680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15051680 Supplier MP Biomedicals, Inc Supplier No. 0215051680
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Triarylmethane dye used for the staining of gram positive bacteria.

TRUSTED_SUSTAINABILITY

Specifications

CAS 633-03-4
Molecular Formula C27H34N2O4S
Color Green
Synonym Basic green l, C.I. 42040
Quantity 100 g
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Physical Form Crystalline Powder
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
Molecular Weight (g/mol) 482.639
PubChem CID 12449
Formula Weight 482.65
Grade Research
Chemical Name or Material Brilliant green
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.